Assay Details
Gene Family ID: | MIPF0000333, mir-652 |
Mature miRNA Sequence: | CAACCCUAGGAGAGGGUGCCAUUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-652-5p | MIMAT0022709 |
| Cricetulus griseus | cgr-miR-652-5p | MIMAT0023991 |
Stem-loop Details
Human
Cricetulus griseus

Genomic Map