Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. X - 110055725 - 110055725 [-] on 110055725 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-652-5p | MIMAT0022709 |
| Human | hsa-miR-652-3p | MIMAT0003322 |
Stem-loop Details
Human

Genomic Map