Assay Details
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | AUCACAUUGCCAGGGAUUACC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-23b-3p | MIMAT0000125 |
| Rat | rno-miR-23b-3p | MIMAT0000793 |
| Anolis carolinensis | aca-miR-23b-3p | MIMAT0021898 |
| Astatotilapia burtoni | abu-miR-23b | MIMAT0042134 |
| Callithrix jacchus | cja-miR-23b | MIMAT0039380 |
| Chicken | gga-miR-23b-3p | MIMAT0001186 |
| View More Rows | ||
| Chrysemys picta | cpi-miR-23b-3p | MIMAT0037656 |
| Cricetulus griseus | cgr-miR-23b-3p | MIMAT0023865 |
| Cyprinus carpio | ccr-miR-23b | MIMAT0026278 |
| Daubentonia madagascariensis | dma-miR-23b | MIMAT0049279 |
| Goat | chi-miR-23b-3p | MIMAT0036089 |
| Horse | eca-miR-23b | MIMAT0013113 |
| Ictalurus punctatus | ipu-miR-23b | MIMAT0029482 |
| Metriaclima zebra | mze-miR-23b | MIMAT0042354 |
| Microcebus murinus | mmr-miR-23b | MIMAT0049228 |
| Monodelphis domestica | mdo-miR-23b-3p | MIMAT0004176 |
| Neolamprologus brichardi | nbr-miR-23b | MIMAT0042535 |
| Nomascus leucogenys | nle-miR-23b | MIMAT0049446 |
| Oreochromis niloticus | oni-miR-23b | MIMAT0042713 |
| Papio hamadryas | pha-miR-23b | MIMAT0049526 |
| Pundamilia nyererei | pny-miR-23b | MIMAT0042924 |
| Rhesus monkey | mml-miR-23b-3p | MIMAT0006165 |
| Taeniopygia guttata | tgu-miR-23-3p | MIMAT0014566 |
| Xenopus laevis | xla-miR-23b-3p | MIMAT0046549 |
| Xenopus tropicalis | xtr-miR-23b | MIMAT0003673 |
Stem-loop Details
Mouse
Rat
Anolis carolinensis
Astatotilapia burtoni
Callithrix jacchus
Chicken
Chrysemys picta
Cricetulus griseus
Cyprinus carpio
Daubentonia madagascariensis
Goat
Horse
Ictalurus punctatus
Metriaclima zebra
Microcebus murinus
Monodelphis domestica
Neolamprologus brichardi
Nomascus leucogenys
Oreochromis niloticus
Papio hamadryas
Pundamilia nyererei
Rhesus monkey
Taeniopygia guttata
Xenopus laevis
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-23b-3p (v21)
Human: hsa-miR-23b (v17)
Mouse: mmu-miR-23b (v17)
Rat: rno-miR-23b (v18)
Aeromonas caviae Ae398: aca-miR-23b (v18)
Gallus gallus: gga-miR-23b (v19)
Macaca mulatta: mml-miR-23b (v19)
Monodelphis domestica: mdo-miR-23b (v19)
Trabulsiella guamensis ATCC 49490: tgu-miR-23 (v18)
Human: hsa-miR-23b (v17)
Mouse: mmu-miR-23b (v17)
Rat: rno-miR-23b (v18)
Aeromonas caviae Ae398: aca-miR-23b (v18)
Gallus gallus: gga-miR-23b (v19)
Macaca mulatta: mml-miR-23b (v19)
Monodelphis domestica: mdo-miR-23b (v19)
Trabulsiella guamensis ATCC 49490: tgu-miR-23 (v18)

Genomic Map