Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 17 - 823073 - 823073 [-] on 823073 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-23b-3p | MIMAT0000793 |
Rat | rno-miR-23b-5p | MIMAT0017099 |
Stem-loop Details
Rat
