Assay Details
Mature miRNA Sequence: | AGAGUAGUAGGUUGCAUAGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-352 | MIMAT0000610 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-352 |
|---|---|
| miRBase Accession Number: | MI0000644 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAGUAGUAGGUUGCAUAGUA |
| Species: | Rat |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AGAGUAGUAGGUUGCAUAGUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-352 | MIMAT0000610 |