Assay Details
Mature miRNA Sequence: | AGAGUAGUAGGUUGCAUAGUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-352 | MIMAT0000610 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-352 |
---|---|
Stem-loop Accession Number: | MI0000644 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGAGUAGUAGGUUGCAUAGUA |
Species: | Rat |
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | AGAGUAGUAGGUUGCAUAGUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-352 | MIMAT0000610 |