Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | UGGCAGUGUAUUGUUAGCUGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-449a | MIMAT0001541 |
| Mouse | mmu-miR-449a-5p | MIMAT0001542 |
| Rat | rno-miR-449a-5p | MIMAT0001543 |
| Bovine | bta-miR-449a | MIMAT0009320 |
| Callithrix jacchus | cja-miR-449a | MIMAT0039495 |
| Cricetulus griseus | cgr-miR-449a | MIMAT0041140 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-449a-5p | MIMAT0047892 |
| Dog | cfa-miR-449a | MIMAT0001544 |
| Guinea pig | cpo-miR-449a-5p | MIMAT0047257 |
| Horse | eca-miR-449a | MIMAT0013102 |
| Monodelphis domestica | mdo-miR-449a-5p | MIMAT0004140 |
| Pongo pygmaeus | ppy-miR-449a | MIMAT0015891 |
| Pteropus alecto | pal-miR-449a-5p | MIMAT0039939 |
| Rabbit | ocu-miR-449a-5p | MIMAT0048453 |
| Rhesus monkey | mml-miR-449a-5p | MIMAT0006330 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Cricetulus griseus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Monodelphis domestica
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-449a (v17)
Rat: rno-miR-449a (v18)
Citrobacter farmeri: cfa-miR-449 (v20)
Macaca mulatta: mml-miR-449a (v19)
Monodelphis domestica: mdo-miR-449 (v19)
Rat: rno-miR-449a (v18)
Citrobacter farmeri: cfa-miR-449 (v20)
Macaca mulatta: mml-miR-449a (v19)
Monodelphis domestica: mdo-miR-449 (v19)

Genomic Map