Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 13 - 113037918 - 113037918 [+] on 113037918 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-449a-5p | MIMAT0001542 |
| Mouse | mmu-miR-449a-3p | MIMAT0017180 |
Stem-loop Details
Mouse

Genomic Map