Assay Details
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GUCCAGUUUUCCCAGGAAUCCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-145-5p | MIMAT0000437 |
| Mouse | mmu-miR-145a-5p | MIMAT0000157 |
| Rat | rno-miR-145-5p | MIMAT0000851 |
| Bovine | bta-miR-145 | MIMAT0003542 |
| Callithrix jacchus | cja-miR-145 | MIMAT0039414 |
| Chrysemys picta | cpi-miR-145-5p | MIMAT0037772 |
| View More Rows | ||
| Columba livia | cli-miR-145-5p | MIMAT0038533 |
| Daubentonia madagascariensis | dma-miR-145 | MIMAT0049256 |
| Dog | cfa-miR-145 | MIMAT0009863 |
| Gadus morhua | gmo-miR-145-5p | MIMAT0044334 |
| Goat | chi-miR-145-5p | MIMAT0035967 |
| Guinea pig | cpo-miR-145-5p | MIMAT0047014 |
| Horse | eca-miR-145 | MIMAT0013064 |
| Ictalurus punctatus | ipu-miR-145 | MIMAT0029422 |
| Microcebus murinus | mmr-miR-145 | MIMAT0049208 |
| Monodelphis domestica | mdo-miR-145-5p | MIMAT0004116 |
| Nomascus leucogenys | nle-miR-145 | MIMAT0049426 |
| Oreochromis niloticus | oni-miR-145 | MIMAT0043395 |
| Otolemur garnettii | oga-miR-145 | MIMAT0049668 |
| Pan paniscus | ppa-miR-145 | MIMAT0049057 |
| Papio hamadryas | pha-miR-145 | MIMAT0049513 |
| Petromyzon marinus | pma-miR-145-5p | MIMAT0019476 |
| Pteropus alecto | pal-miR-145-5p | MIMAT0040158 |
| Python bivittatus | pbv-miR-145-5p | MIMAT0038959 |
| Rabbit | ocu-miR-145-5p | MIMAT0048251 |
| Salmo salar | ssa-miR-145-5p | MIMAT0032369 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Chrysemys picta
Columba livia
Daubentonia madagascariensis
Dog
Gadus morhua
Goat
Guinea pig
Horse
Ictalurus punctatus
Microcebus murinus
Monodelphis domestica
Nomascus leucogenys
Oreochromis niloticus
Otolemur garnettii
Pan paniscus
Papio hamadryas
Petromyzon marinus
Pteropus alecto
Python bivittatus
Rabbit
Salmo salar
More Information
Related Products
miRBase Alias:
Human: hsa-miR-145 (v17)
Mouse: mmu-miR-145-5p (v18)
Mouse: mmu-miR-145 (v17)
Rat: rno-miR-145 (v18)
Monodelphis domestica: mdo-miR-145 (v19)
Phocea massiliensis: pma-miR-145 (v18)
Mouse: mmu-miR-145-5p (v18)
Mouse: mmu-miR-145 (v17)
Rat: rno-miR-145 (v18)
Monodelphis domestica: mdo-miR-145 (v19)
Phocea massiliensis: pma-miR-145 (v18)

Genomic Map