Assay Details
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GUCCAGUUUUCCCAGGAAUCCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-145-5p | MIMAT0000437 |
| Mouse | mmu-miR-145a-5p | MIMAT0000157 |
| Rat | rno-miR-145-5p | MIMAT0000851 |
| Bovine | bta-miR-145 | MIMAT0003542 |
| Callithrix jacchus | cja-miR-145 | MIMAT0039414 |
| Chrysemys picta | cpi-miR-145-5p | MIMAT0037772 |
| View More Rows | ||
| Columba livia | cli-miR-145-5p | MIMAT0038533 |
| Daubentonia madagascariensis | dma-miR-145 | MIMAT0049256 |
| Dog | cfa-miR-145 | MIMAT0009863 |
| Gadus morhua | gmo-miR-145-5p | MIMAT0044334 |
| Goat | chi-miR-145-5p | MIMAT0035967 |
| Guinea pig | cpo-miR-145-5p | MIMAT0047014 |
| Horse | eca-miR-145 | MIMAT0013064 |
| Ictalurus punctatus | ipu-miR-145 | MIMAT0029422 |
| Microcebus murinus | mmr-miR-145 | MIMAT0049208 |
| Monodelphis domestica | mdo-miR-145-5p | MIMAT0004116 |
| Nomascus leucogenys | nle-miR-145 | MIMAT0049426 |
| Oreochromis niloticus | oni-miR-145 | MIMAT0043395 |
| Otolemur garnettii | oga-miR-145 | MIMAT0049668 |
| Pan paniscus | ppa-miR-145 | MIMAT0049057 |
| Papio hamadryas | pha-miR-145 | MIMAT0049513 |
| Petromyzon marinus | pma-miR-145-5p | MIMAT0019476 |
| Pteropus alecto | pal-miR-145-5p | MIMAT0040158 |
| Python bivittatus | pbv-miR-145-5p | MIMAT0038959 |
| Rabbit | ocu-miR-145-5p | MIMAT0048251 |
| Salmo salar | ssa-miR-145-5p | MIMAT0032369 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-145 |
| Stem-loop Accession # | MI0000461 |
| Stem-loop Sequence | CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU |
| Chromosome Location | Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000437
miRBase ID: hsa-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-145a |
| Stem-loop Accession # | MI0000169 |
| Stem-loop Sequence | CUCACGGUCCAGUUUUCCCAGGAAUCCCUUGGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAG |
| Chromosome Location | Chr. 18 - 61647825 - 61647894 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000157
miRBase ID: mmu-miR-145a-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: Chr. 18 - 61647825 - 61647894 [-] on Build GRCm38 |
| Stem-loop ID | rno-mir-145 |
| Stem-loop Accession # | MI0000918 |
| Stem-loop Sequence | CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUGGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGCU |
| Chromosome Location | Chr. 18 - 56969907 - 56969994 [-] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000851
miRBase ID: rno-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: Chr. 18 - 56969907 - 56969994 [-] on Build Rnor_6.0 |
| Stem-loop ID | bta-mir-145 |
| Stem-loop Accession # | MI0004756 |
| Stem-loop Sequence | CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003542
miRBase ID: bta-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | cja-mir-145 |
| Stem-loop Accession # | MI0031934 |
| Stem-loop Sequence | ACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGACGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039414
miRBase ID: cja-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | cpi-mir-145 |
| Stem-loop Accession # | MI0029488 |
| Stem-loop Sequence | GUGUCCUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGGCACUAUGACGGGGAUUCCUGGAAAUACUGUUCUUGAGGCCAUGGCUCAGCAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037772
miRBase ID: cpi-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | cli-mir-145 |
| Stem-loop Accession # | MI0029979 |
| Stem-loop Sequence | CCGUGCCCUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGGCGCUACGUUGGGGAUUCCUGGAAAUACUGUUCUUGAGGCCACGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038533
miRBase ID: cli-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | dma-mir-145 |
| Stem-loop Accession # | MI0039984 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049256
miRBase ID: dma-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-145 |
| Stem-loop Accession # | MI0010359 |
| Stem-loop Sequence | UGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009863
miRBase ID: cfa-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-145 |
| Stem-loop Accession # | MI0036228 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUGUUGAACAUGAAAGCGGGAUUCCUGGAAAUACUGUUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044334
miRBase ID: gmo-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | chi-mir-145 |
| Stem-loop Accession # | MI0030631 |
| Stem-loop Sequence | CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0035967
miRBase ID: chi-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-145 |
| Stem-loop Accession # | MI0038652 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUGGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047014
miRBase ID: cpo-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | eca-mir-145 |
| Stem-loop Accession # | MI0012808 |
| Stem-loop Sequence | CUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013064
miRBase ID: eca-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | ipu-mir-145 |
| Stem-loop Accession # | MI0024511 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCGAAAGGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029422
miRBase ID: ipu-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | mmr-mir-145 |
| Stem-loop Accession # | MI0039931 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049208
miRBase ID: mmr-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | mdo-mir-145 |
| Stem-loop Accession # | MI0005305 |
| Stem-loop Sequence | CUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004116
miRBase ID: mdo-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | nle-mir-145 |
| Stem-loop Accession # | MI0040168 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049426
miRBase ID: nle-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | oni-mir-145 |
| Stem-loop Accession # | MI0035432 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUGACCUAUCAGAAAGGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0043395
miRBase ID: oni-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | oga-mir-145 |
| Stem-loop Accession # | MI0040451 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049668
miRBase ID: oga-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | ppa-mir-145 |
| Stem-loop Accession # | MI0039765 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049057
miRBase ID: ppa-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | pha-mir-145 |
| Stem-loop Accession # | MI0040278 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAAAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049513
miRBase ID: pha-miR-145 Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | pma-mir-145 |
| Stem-loop Accession # | MI0017080 |
| Stem-loop Sequence | UGCGUGGGCAUGGUCCAGUUUUCCCAGGAAUCCCUGUGCCCUUUUCCCGGGAUUCCAGGAAUAACUGUCCAAGCACACGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0019476
miRBase ID: pma-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | pal-mir-145 |
| Stem-loop Accession # | MI0032524 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040158
miRBase ID: pal-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-145 |
| Stem-loop Accession # | MI0030243 |
| Stem-loop Sequence | ACUAUGUCCUCGGGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAAGGGGAUUCCUGGAAAUACUGUUCUUGGGGUCAUGGCUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038959
miRBase ID: pbv-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-145 |
| Stem-loop Accession # | MI0039303 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048251
miRBase ID: ocu-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-145-1 |
| Stem-loop Accession # | MI0026505 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCAACAACGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032369
miRBase ID: ssa-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-145-2 |
| Stem-loop Accession # | MI0026506 |
| Stem-loop Sequence | GUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCACAAGGGGGAUUCCUGGAAAUACUGUUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032369
miRBase ID: ssa-miR-145-5p Mature miRNA Sequence: GUCCAGUUUUCCCAGGAAUCCCU Chromosome Location: NA |
More Information
Related Products

Genomic Map