Assay Details
Gene Family ID: | MIPF0000121, mir-202 |
Mature miRNA Sequence: | UUCCUAUGCAUAUACUUCUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-202-5p | MIMAT0004546 |
| Bovine | bta-miR-202 | MIMAT0009259 |
| Callithrix jacchus | cja-miR-202 | MIMAT0039523 |
| Columba livia | cli-miR-202-5p | MIMAT0038579 |
| Cricetulus griseus | cgr-miR-202 | MIMAT0041155 |
| Goat | chi-miR-202-5p | MIMAT0036048 |
| View More Rows | ||
| Guinea pig | cpo-miR-202-5p | MIMAT0047095 |
| Pig | ssc-miR-202-5p | MIMAT0013948 |
| Pteropus alecto | pal-miR-202-5p | MIMAT0039862 |
Stem-loop Details
Mouse
Bovine
Callithrix jacchus
Columba livia
Cricetulus griseus
Goat
Guinea pig
Pig
Pteropus alecto
More Information
Related Products
miRBase Alias:
Staphylococcus schleiferi: ssc-miR-202 (v18)

Genomic Map