Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 7 - 139957489 - 139957489 [-] on 139957489 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-202-3p | MIMAT0000235 |
| Mouse | mmu-miR-202-5p | MIMAT0004546 |
Stem-loop Details
Mouse

Genomic Map