Assay Details
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | CCAAUAUUGGCUGUGCUGCUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-195-3p | MIMAT0004615 |
| Mouse | mmu-miR-195a-3p | MIMAT0017000 |
Stem-loop Details
Human
Mouse
More Information
Related Products
miRBase Alias:
Human: hsa-miR-195* (v17)
Mouse: mmu-miR-195* (v17)
Mouse: mmu-miR-195-3p (v18)
Mouse: mmu-miR-195* (v17)
Mouse: mmu-miR-195-3p (v18)

Genomic Map