Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 11 - 70235444 - 70235444 [-] on 70235444 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-195a-5p | MIMAT0000225 |
| Mouse | mmu-miR-195a-3p | MIMAT0017000 |
Stem-loop Details
Mouse

Genomic Map