Assay Details
Gene Family ID: | MIPF0000938, mir-2355 |
Mature miRNA Sequence: | AUCCCCAGAUACAAUGGACAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2355-5p | MIMAT0016895 |
Pongo pygmaeus | ppy-miR-2355 | MIMAT0049334 |
Stem-loop Details
Human
Pongo pygmaeus
