Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AAGCUUUUUGCUCGCGUUAUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-208b-5p | MIMAT0017280 |
| Dasypus novemcinctus | dno-miR-208b-5p | MIMAT0047752 |
Stem-loop Details
Mouse
Dasypus novemcinctus

Genomic Map