Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 14 - 54975496 - 54975496 [+] on 54975496 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-208b-3p | MIMAT0004939 |
| Mouse | mmu-miR-208b-5p | MIMAT0017280 |
Stem-loop Details
Mouse

Genomic Map