Assay Details
Gene Family ID: | MIPF0000669, mir-1247 |
Mature miRNA Sequence: | CGGGAACGUCGAGACUGGAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1247-3p | MIMAT0014801 |
| Rat | rno-miR-1247-3p | MIMAT0035722 |
| Callithrix jacchus | cja-miR-1247 | MIMAT0039403 |
| Otolemur garnettii | oga-miR-1247 | MIMAT0049672 |
| Rhesus monkey | mml-miR-1247-3p | MIMAT0028359 |
Stem-loop Details
Mouse
Rat
Callithrix jacchus
Otolemur garnettii
Rhesus monkey

Genomic Map