Assay Details
Amplicon Length: | 105 |
Chromosome Location: | Chr. 12 - 110277901 - 110277901 [-] on 110277901 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1247-5p | MIMAT0014800 |
| Mouse | mmu-miR-1247-3p | MIMAT0014801 |
Stem-loop Details
Mouse

Genomic Map