Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAAUCUACUACCUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3596d | MIMAT0017823 |
Artibeus jamaicensis | aja-miR-3596 | MIMAT0031134 |
Stem-loop Details
Rat
Artibeus jamaicensis
