Assay Details
Gene Family ID: | MIPF0000531, mir-1306 |
Mature miRNA Sequence: | CACCACCUCCCCUGCAAACGUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1306-5p | MIMAT0019136 |
| Ophiophagus hannah | oha-miR-1306-5p | MIMAT0036685 |
Stem-loop Details
Mouse
Ophiophagus hannah

Genomic Map