Assay Details
Amplicon Length: | 107 |
Chromosome Location: | Chr. 16 - 18284680 - 18284680 [+] on 18284680 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1306-5p | MIMAT0019136 |
| Mouse | mmu-miR-1306-3p | MIMAT0009411 |
Stem-loop Details
Mouse

Genomic Map