Assay Details
Gene Family ID: | MIPF0000598, mir-1298 |
Mature miRNA Sequence: | CAUCUGGGCAACUGACUGAAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1298-3p | MIMAT0026641 |
| Callithrix jacchus | cja-miR-1298 | MIMAT0039398 |
Stem-loop Details
Human
Callithrix jacchus

Genomic Map