Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. X - 114715644 - 114715644 [-] on 114715644 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1298-3p | MIMAT0026641 |
| Human | hsa-miR-1298-5p | MIMAT0005800 |
Stem-loop Details
Human

Genomic Map