Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 19 - 22750305 - 22750305 [+] on 22750305 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-204-5p | MIMAT0000237 |
| Mouse | mmu-miR-204-3p | MIMAT0017002 |
Stem-loop Details
Mouse

Genomic Map