Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. X - 151912148 - 151912148 [-] on 151912148 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-let-7f-5p | MIMAT0000525 |
Mouse | mmu-let-7f-2-3p | MIMAT0017017 |
Stem-loop Details
Mouse
