Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAGUCUACUGUCUUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-let-7f-2-3p | MIMAT0017017 |
Rat | rno-let-7f-2-3p | MIMAT0017090 |
Anolis carolinensis | aca-let-7a-3p | MIMAT0021693 |
Ornithorhynchus anatinus | oan-let-7f-2-3p | MIMAT0009171 |
Python bivittatus | pbv-let-7a-3p | MIMAT0038794 |
Stem-loop Details
Mouse
Rat
Anolis carolinensis
Ornithorhynchus anatinus
Python bivittatus

More Information
Related Products
miRBase Alias:
Mouse: mmu-let-7f-2* (v17)
Rat: rno-let-7f-2* (v18)
Aeromonas caviae Ae398: aca-let-7a* (v18)
Ochrobactrum anthropi: oan-let-7f-2* (v18)
Rat: rno-let-7f-2* (v18)
Aeromonas caviae Ae398: aca-let-7a* (v18)
Ochrobactrum anthropi: oan-let-7f-2* (v18)
