Assay Details
Amplicon Length: | 58 |
Chromosome Location: | Chr. 4 - 88910358 - 88910358 [+] on 88910358 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-31-3p | MIMAT0004634 |
Mouse | mmu-miR-31-5p | MIMAT0000538 |
Stem-loop Details
Mouse
