Assay Details
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | AGGCAAGAUGCUGGCAUAGCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-31-5p | MIMAT0000538 | |
| Rat | rno-miR-31a-5p | MIMAT0000810 | |
| Callithrix jacchus | cja-miR-31 | MIMAT0039372 | |
| Cricetulus griseus | cgr-miR-31-5p | MIMAT0023902 | |
| Daubentonia madagascariensis | dma-miR-31 | MIMAT0049270 | |
| Microcebus murinus | mmr-miR-31 | MIMAT0049214 | |
| View More Rows | |||
| Nomascus leucogenys | nle-miR-31 | MIMAT0049444 | |
| Otolemur garnettii | oga-miR-31 | MIMAT0049629 | |
| Papio hamadryas | pha-miR-31 | MIMAT0049531 | |
| Pig | ssc-miR-31 | MIMAT0025360 | |
Stem-loop Details
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Daubentonia madagascariensis
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Pig
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-31 (v17)
Rat: rno-miR-31 (v18)
Rat: rno-miR-31 (v18)

Genomic Map