Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 11 - 70012347 - 70012347 [-] on 70012347 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-324-3p | MIMAT0000556 |
| Mouse | mmu-miR-324-5p | MIMAT0000555 |
Stem-loop Details
Mouse

Genomic Map