Assay Details
Gene Family ID: | MIPF0000165, mir-324 |
Mature miRNA Sequence: | CGCAUCCCCUAGGGCAUUGGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-324-5p | MIMAT0000555 |
| Rat | rno-miR-324-5p | MIMAT0000553 |
| Bovine | bta-miR-324 | MIMAT0009285 |
| Callithrix jacchus | cja-miR-324 | MIMAT0039402 |
| Dasypus novemcinctus | dno-miR-324-5p | MIMAT0047803 |
| Dog | cfa-miR-324 | MIMAT0009896 |
| View More Rows | ||
| Goat | chi-miR-324-5p | MIMAT0036135 |
| Guinea pig | cpo-miR-324-5p | MIMAT0047158 |
| Horse | eca-miR-324-5p | MIMAT0013033 |
| Nomascus leucogenys | nle-miR-324 | MIMAT0049336 |
| Pig | ssc-miR-324 | MIMAT0013957 |
| Pongo pygmaeus | ppy-miR-324-5p | MIMAT0015827 |
| Rabbit | ocu-miR-324-5p | MIMAT0048370 |
| Rhesus monkey | mml-miR-324-5p | MIMAT0006266 |
| Tupaia chinensis | tch-miR-324-5p | MIMAT0036596 |
Stem-loop Details
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Nomascus leucogenys
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Tupaia chinensis

Genomic Map