Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. 12 - 109712008 - 109712008 [+] on 109712008 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-380-5p | MIMAT0000744 |
| Mouse | mmu-miR-380-3p | MIMAT0000745 |
Stem-loop Details
Mouse

Genomic Map