Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | AUGGUUGACCAUAGAACAUGCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-380-5p | MIMAT0000744 |
| Rat | rno-miR-380-5p | MIMAT0005308 |
| Dasypus novemcinctus | dno-miR-380-5p | MIMAT0047862 |
| Guinea pig | cpo-miR-380-5p | MIMAT0047223 |
| Rhesus monkey | mml-miR-380-5p | MIMAT0026866 |
Stem-loop Details
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Rhesus monkey

Genomic Map