Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 1 - 20683074 - 20683074 [-] on 20683074 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-133b-3p | MIMAT0000769 |
| Mouse | mmu-miR-133b-5p | MIMAT0017083 |
Stem-loop Details
Mouse

Genomic Map