Assay Details
Gene Family ID: | MIPF0000029, mir-133 |
Mature miRNA Sequence: | UUUGGUCCCCUUCAACCAGCUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-133b | MIMAT0000770 | |
| Mouse | mmu-miR-133b-3p | MIMAT0000769 | |
| Rat | rno-miR-133b-3p | MIMAT0003126 | |
| Alligator mississippiensis | ami-miR-133b-3p | MIMAT0038148 | |
| Bovine | bta-miR-133b | MIMAT0009226 | |
| Columba livia | cli-miR-133b-3p | MIMAT0038512 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-133b-3p | MIMAT0047636 | |
| Dog | cfa-miR-133b | MIMAT0009835 | |
| Guinea pig | cpo-miR-133b-3p | MIMAT0046989 | |
| Horse | eca-miR-133b | MIMAT0013097 | |
| Ornithorhynchus anatinus | oan-miR-133b-3p | MIMAT0006997 | |
| Pan troglodytes | ptr-miR-133b | MIMAT0008030 | |
| Pongo pygmaeus | ppy-miR-133b | MIMAT0015756 | |
| Python bivittatus | pbv-miR-133b-3p | MIMAT0038939 | |
| Rabbit | ocu-miR-133b-3p | MIMAT0048224 | |
| Rhesus monkey | mml-miR-133b-3p | MIMAT0006189 | |
| Zebrafish | dre-miR-133b-3p | MIMAT0001831 | |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Bovine
Columba livia
Dasypus novemcinctus
Dog
Guinea pig
Horse
Ornithorhynchus anatinus
Pan troglodytes
Pongo pygmaeus
Python bivittatus
Rabbit
Rhesus monkey
Zebrafish
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Mouse: mmu-miR-133b (v17)
Rat: rno-miR-133b (v18)
Danio rerio: dre-miR-133b (v18)
Macaca mulatta: mml-miR-133b (v19)
Ochrobactrum anthropi: oan-miR-133b (v18)
Rat: rno-miR-133b (v18)
Danio rerio: dre-miR-133b (v18)
Macaca mulatta: mml-miR-133b (v19)
Ochrobactrum anthropi: oan-miR-133b (v18)

Genomic Map