Assay Details
Amplicon Length: | 115 |
Chromosome Location: | Chr. X - 66792393 - 66792393 [-] on 66792393 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-471-5p | MIMAT0002112 |
| Mouse | mmu-miR-471-3p | MIMAT0017195 |
Stem-loop Details
Mouse

Genomic Map