Assay Details
Gene Family ID: | MIPF0000423, mir-471 |
Mature miRNA Sequence: | UACGUAGUAUAGUGCUUUUCAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-471-5p | MIMAT0002112 |
| Rat | rno-miR-471-5p | MIMAT0005318 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-471 (v15)
Rat: rno-miR-471 (v18)
Rat: rno-miR-471 (v18)

Genomic Map