Assay Details
Amplicon Length: | 101 |
Chromosome Location: | Chr. 7 - 3219784 - 3219784 [+] on 3219784 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-291b-5p | MIMAT0003189 |
| Mouse | mmu-miR-291b-3p | MIMAT0003190 |
| Mouse | mmu-miR-292a-3p | MIMAT0000370 |
| Mouse | mmu-miR-292a-5p | MIMAT0000369 |
| Mouse | mmu-miR-292b-5p | MIMAT0029864 |
| Mouse | mmu-miR-292b-3p | MIMAT0029865 |
Stem-loop Details
Mouse

Genomic Map