Assay Details
Gene Family ID: | MIPF0000068, mir-290 |
Mature miRNA Sequence: | ACUCAAACUGGGGGCUCUUUUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-292a-5p | MIMAT0000369 |
| Rat | rno-miR-292-5p | MIMAT0000896 |
Stem-loop Details
Mouse
Rat

Genomic Map