Assay Details
Amplicon Length: | 53 |
Chromosome Location: | Chr. 12 - 109586716 - 109586716 [-] on 109586716 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-665-5p | MIMAT0017238 |
| Mouse | mmu-miR-665-3p | MIMAT0003733 |
Stem-loop Details
Mouse

Genomic Map