Assay Details
Gene Family ID: | MIPF0000404, mir-665 |
Mature miRNA Sequence: | ACCAGGAGGCUGAGGUCCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-665-3p | MIMAT0003733 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-665 (v17)
Rat: rno-miR-665 (v15)
Rat: rno-miR-665 (v15)

Genomic Map