Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 7 - 3218463 - 3218463 [-] on 3218463 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-290a-5p | MIMAT0000366 |
| Mouse | mmu-miR-290a-3p | MIMAT0004572 |
| Mouse | mmu-miR-290b-5p | MIMAT0029900 |
| Mouse | mmu-miR-290b-3p | MIMAT0029901 |
| Mouse | mmu-miR-291a-3p | MIMAT0000368 |
| Mouse | mmu-miR-291a-5p | MIMAT0000367 |
Stem-loop Details
Mouse

Genomic Map