Assay Details
Gene Family ID: | MIPF0000068, mir-290 |
Mature miRNA Sequence: | CAUCAAAGUGGAGGCCCUCUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-291a-5p | MIMAT0000367 |
| Rat | rno-miR-291a-5p | MIMAT0000894 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-291-5p (v7.1)
Rat: rno-miR-291-5p (v9.2)
Rat: rno-miR-291-5p (v9.2)

Genomic Map