Assay Details
Amplicon Length: | 102 |
Chromosome Location: | Chr. X - 53053830 - 53053830 [-] on 53053830 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-503-3p | MIMAT0004790 |
Mouse | mmu-miR-503-5p | MIMAT0003188 |
Stem-loop Details
Mouse
