Assay Details
Gene Family ID: | MIPF0000183, mir-503 |
Mature miRNA Sequence: | UAGCAGCGGGAACAGUACUGCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-503-5p | MIMAT0003188 |
| Rat | rno-miR-503-5p | MIMAT0003213 |
| Dasypus novemcinctus | dno-miR-503-5p | MIMAT0047944 |
| Guinea pig | cpo-miR-503-5p | MIMAT0047307 |
| Horse | eca-miR-503 | MIMAT0013227 |
| Pig | ssc-miR-503 | MIMAT0010189 |
Stem-loop Details
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Horse
Pig

Genomic Map