Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 1 - 185242825 - 185242825 [-] on 185242825 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-664-5p | MIMAT0017353 |
| Mouse | mmu-miR-664-3p | MIMAT0012774 |
Stem-loop Details
Mouse

Genomic Map