Assay Details
Gene Family ID: | MIPF0000300, mir-664 |
Mature miRNA Sequence: | UAUUCAUUUACUCCCCAGCCUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-664-3p | MIMAT0012774 |
Rat | rno-miR-664-3p | MIMAT0003382 |
Stem-loop Details
Mouse
Rat

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-664 (v17)
Rat: rno-miR-664 (v18)
Rat: rno-miR-664 (v18)
