Assay Details
Gene Family ID: | MIPF0000244, mir-351 |
Mature miRNA Sequence: | UCCCUGAGGAGCCCUUUGAGCCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-351-5p | MIMAT0000609 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-351 (v17)
Rat: rno-miR-351 (v9.2)
Rat: rno-miR-351 (v9.2)

Genomic Map