Assay Details
Amplicon Length: | 53 |
Chromosome Location: | Chr. X - 53052956 - 53052956 [+] on 53052956 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-351-3p | MIMAT0017042 |
| Mouse | mmu-miR-351-5p | MIMAT0000609 |
Stem-loop Details
Mouse

Genomic Map