Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CCUCUAGAUGGAAGCACUGUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-517-5p | MIMAT0002851 |
| Gorilla gorilla | ggo-miR-517c-5p | MIMAT0036432 |
| Pan troglodytes | ptr-miR-517b | MIMAT0008185 |
| Pongo pygmaeus | ppy-miR-517c-5p | MIMAT0036447 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus

Genomic Map